Merck
Metabolic Induction of Trained Immunity through the Mevalonate Pathway.
Cell (2018-01-13)
Siroon Bekkering, Rob J W Arts, Boris Novakovic, Ioannis Kourtzelis, Charlotte D C C van der Heijden, Yang Li, Calin D Popa, Rob Ter Horst, Julia van Tuijl, Romana T Netea-Maier, Frank L van de Veerdonk, Triantafyllos Chavakis, Leo A B Joosten, Jos W M van der Meer, Henk Stunnenberg, Niels P Riksen, Mihai G Netea
ZUSAMMENFASSUNG
Innate immune cells can develop long-term memory after stimulation by microbial products during infections or vaccinations. Here, we report that metabolic signals can induce trained immunity. Pharmacological and genetic experiments reveal that activation of the cholesterol synthesis pathway, but not the synthesis of cholesterol itself, is essential for training of myeloid cells. Rather, the metabolite mevalonate is the mediator of training via activation of IGF1-R and mTOR and subsequent histone modifications in inflammatory pathways. Statins, which block mevalonate generation, prevent trained immunity induction. Furthermore, monocytes of patients with hyper immunoglobulin D syndrome (HIDS), who are mevalonate kinase deficient and accumulate mevalonate, have a constitutive trained immunity phenotype at both immunological and epigenetic levels, which could explain the attacks of sterile inflammation that these patients experience. Unraveling the role of mevalonate in trained immunity contributes to our understanding of the pathophysiology of HIDS and identifies novel therapeutic targets for clinical conditions with excessive activation of trained immunity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Aktin, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Type II-A, ≥60% (SDS-PAGE), >= 60 Kunitz units/mg protein
Sigma-Aldrich
Fluvastatin sodium hydrate, ≥98% (HPLC)
Sigma-Aldrich
Picropodophyllotoxin, ≥96% (HPLC), powder
Sigma-Aldrich
GSK3787, ≥98% (HPLC), white to off-white, powder
Sigma-Aldrich
6-Fluoromevalonate, ≥90% (GC), viscous liquid