Merck
  • Fluorescence-activated sorting of rat hepatocytes based on their mixed function oxidase activities towards diethoxyfluorescein.
Fluorescence-activated sorting of rat hepatocytes based on their mixed function oxidase activities towards diethoxyfluorescein.
The Biochemical journal (1987-10-01)
I N White, M L Green, R F Legg
ZUSAMMENFASSUNG
The formation of ethoxyfluorescein and fluorescein from diethoxyfluorescein by isolated rat hepatocytes has been used as a basis for separating such cells dependent on their mixed function oxidase activities by fluorescence-activated flow cytometry. Five equal fractions defined by computer-generated regions were isolated. Non-viable cells with low fluorescence (region 1) represented 10-15% of the population, while the remainder with higher mixed function oxidase activities (regions 2-5), were greater than 95% viable by Trypan Blue exclusion. In region 1, 30% of the viable cells were binucleate, 67% diploid while in region 5, 13% were binucleate and 69% tetraploid. At 3 h after sorting, following attachment to glass coverslips, exposure of cells to methyl methanesulphonate, retrorsine or norethindrone resulted in unscheduled DNA synthesis which was 2-fold higher in the tetraploid-rich region 5, while aflatoxin B1, benzo[a]pyrene or 2-acetylaminofluorene caused a 5-fold increase in unscheduled DNA synthesis in these cells, relative to the diploid-rich hepatocytes in region 2.