Merck
  • A Reaction-Based Novel Fluorescent Probe for Detection of Hydrogen Sulfide and Its Application in Wine.
A Reaction-Based Novel Fluorescent Probe for Detection of Hydrogen Sulfide and Its Application in Wine.
Journal of food science (2017-12-16)
Hao Wang, Jialin Wang, Shaoxiang Yang, Hongyu Tian, Baoguo Sun, Yongguo Liu
ZUSAMMENFASSUNG
A new reaction-based fluorescent probe 6-cyanonaphthalen-2-yl-2,4- dinitrobenzenesulfonate (probe 1) was designed and synthesized for detection of hydrogen sulfide (H Fluorescent probe 1 could be employed as a visible sensor for H
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-Cystein, technical grade
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
6-Cyano-2-naphthol, 97%