Merck
Plasma membrane Ca
The Journal of physiology (2017-10-13)
Yanwen Wang, Claire Wilson, Elizabeth J Cartwright, Ming Lei
ZUSAMMENFASSUNG
The role of plasma membrane Ca To determine the role of plasma membrane Ca
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade