Merck
  • Supramolecular Hydrogel Formation Based on Inclusion Complexation Between Poly (ethylene glycol)?Modified Chitosan and ??Cyclodextrin.
Supramolecular Hydrogel Formation Based on Inclusion Complexation Between Poly (ethylene glycol)?Modified Chitosan and ??Cyclodextrin.
Macromolecular Bioscience (2004)
Huh K