Merck
Generation of double-knockout embryonic stem cells.
Methods in molecular biology (Clifton, N.J.) (2009-03-07)
Eva Wielders, Marleen Dekker, Hein Te Riele
ZUSAMMENFASSUNG
Gene inactivation in mouse embryonic stem (ES) cells usually affects a single allele that is subsequently transmitted to the mouse germline. Upon breeding to homozygosity the consequences of complete gene ablation can be studied in the context of the complete organism. In many cases, it can be useful to study the consequences of gene ablation already in ES cells, for example, when a cellular phenotype is expected. This requires both alleles of a gene to be disrupted. Besides consecutive targeting by using different selectable marker genes, homozygosity for gene disruption can also be obtained by selecting cells for duplication of (part of) the chromosome carrying the targeted allele with concomitant loss of the wild-type allele.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Gelatine aus Schweinehaut, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
ESGRO® Rekombinantes Maus-LIF-Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^7 units/ml.
Sigma-Aldrich
Puromycin -dihydrochlorid aus Streptomyces alboniger, ≥98% (HPLC), powder