Merck
  • Different patterns of neuronal activation and neurodegeneration in the thalamus and cortex of epilepsy-resistant Proechimys rats versus Wistar rats after pilocarpine-induced protracted seizures.
Different patterns of neuronal activation and neurodegeneration in the thalamus and cortex of epilepsy-resistant Proechimys rats versus Wistar rats after pilocarpine-induced protracted seizures.
Epilepsia (2009-02-18)
Anna Andrioli, Paolo F Fabene, Roberto Spreafico, Esper A Cavalheiro, Marina Bentivoglio
ZUSAMMENFASSUNG
To analyze cellular mechanisms of limbic-seizure suppression, the response to pilocarpine-induced seizures was investigated in cortex and thalamus, comparing epilepsy-resistant rats Proechimys guyannensis with Wistar rats. Fos immunoreactivity revealing neuronal activation, and degenerating neurons labeled by Fluoro-Jade B (FJB) histochemistry were analyzed on the first day after onset of seizures lasting 3 h. Subpopulations of gamma-aminobutyric acid (GABA)ergic cells were characterized with double Fos-parvalbumin immunohistochemistry. In both cortex and thalamus, degenerating neurons were much fewer in Proechimys than Wistar rats. Fos persisted at high levels at 24 h only in the Proechimys thalamus and cortex, especially in layer VI where corticothalamic neurons reside. In the parietal cortex, about 50% of parvalbumin-containing interneurons at 8 h, and 10-20% at 24 h, were Fos-positive in Wistar rats, but in Proechimys, Fos was expressed in almost all parvalbumin-containing interneurons at 8 h and dropped at 24 h. Fos positivity in cingulate cortex interneurons was similar in both species. In the Wistar rat thalamus, Fos was induced in medial and midline nuclei up to 8 h, when <30% of reticular nucleus cells were Fos-positive, and then decreased, with no relationship with cell loss, evaluated in Nissl-stained sections. In Proechimys, almost all reticular nucleus neurons were Fos-positive at 24 h. At variance with laboratory rats, pilocarpine-induced protracted seizures elicit in Proechimys limited neuronal death, and marked and long-lasting Fos induction in excitatory and inhibitory cortical and thalamic cell subsets. The findings implicate intrathalamic and intracortical regulation, and circuits linking thalamus and cortex in limbic seizure suppression leading to epilepsy resistance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pilocarpin -nitrat (Salz), ≥98% (HPLC)