Merck
Congener specific analysis of polychlorinated biphenyls (PCBs) in serum using GC/MSD.
Chemosphere (1995-05-01)
U S Gill, H M Schwartz, B Wheatley
ZUSAMMENFASSUNG
A new congener specific method has been developed for extraction and quantitative determination of PCB contaminants of serum. After liquid-liquid extraction and chromatographic fractionation with Carbopack-C/florisil column, the analytes were identified and quantified utilizing gas chromatography/mass spectrometry (GC/MSD) using selected ion monitoring (SIM) technique. Recoveries of 35 non-planar PCB congeners (#18, 28, 31, 44, 49, 52, 60, 66, 74, 82, 87, 99, 101, 105, 110, 114, 118, 128, 138, 141, 146, 151, 153, 156, 157, 158, 167, 170, 171, 180, 183, 185, 187, 189 and 194) and 5 non-ortho (coplanar) congeners (#37, 77, 81, 126 and 169) from spiked serum were between 55-115%.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
FLM Carbopack X TD-Röhrchen aus deaktiviertem Edelstahl, preconditioned, pkg of 10 tubes, O.D. × L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm)
Supelco
Carbopack Adsorbens, matrix Carbopack B, 60-80 mesh, bottle of 10 g
Supelco
Carbopack Adsorbens, matrix Carbopack C, 80-100 mesh, bottle of 10 g
Supelco
Carbopack Adsorbens, matrix Carbopack C, 60-80 mesh, bottle of 10 g
Supelco
Carbopack Adsorbens, matrix Carbopack X, 40-60 mesh, bottle of 10 g
Supelco
Carbopack Adsorbens, matrix Carbopack X, 60-80 mesh, bottle of 10 g
Supelco
Carbopack B, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
Supelco
Carbopack X, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbopack B, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., unconditioned, pkg of 10 ea, matrix Carbopack
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea