Merck
[Determination of prometryne residue in shellfish by gas chromatography].
Wei sheng yan jiu = Journal of hygiene research (2014-12-03)
Yeping Song, Wanli Zong, Zhongfei Yu, Xincai Liu, Ling Xiao, Yan Wang
ZUSAMMENFASSUNG
To develop a method of gas chromatography (GC) for determining residues of promtryne in shellfish. The sample was extracted with ethyl acetate, and cleaned-up with Envi-Carb SPE cartridge, alumina-N SPE cartridge, and determined by GC-FPD with DB - 1701 capillary chromatographic column (30 m x0.53 .mm x 0.5µm). Good linear was obtained in the concentration range of 0.10-0.96µg/ml with a correlation coefficient of 0. 999. The limit of detection (LOD) was 0.0037 mg/kg and limit of quantity (LOQ) was 0.010 mg/kg. The average recoveries was 83. 7% - 102. 0% and relative standard deviations (RSD) was 0. 54% - 8.14% The method is simple, fast and credible, so it can be applied to determination of prometryne in shellfish.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS