Merck
  • α-Amino-β-carboxymuconate-ε-semialdehyde Decarboxylase (ACMSD) Inhibitors as Novel Modulators of De Novo Nicotinamide Adenine Dinucleotide (NAD
α-Amino-β-carboxymuconate-ε-semialdehyde Decarboxylase (ACMSD) Inhibitors as Novel Modulators of De Novo Nicotinamide Adenine Dinucleotide (NAD
Journal of medicinal chemistry (2018-01-19)
Roberto Pellicciari, Paride Liscio, Nicola Giacchè, Francesca De Franco, Andrea Carotti, Janet Robertson, Lucia Cialabrini, Elena Katsyuba, Nadia Raffaelli, Johan Auwerx
ZUSAMMENFASSUNG
NAD
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Discovery® Cyano HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® Cyano Supelguard Kartusche, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® Cyano HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4 mm