Merck
P3 SAR exploration of biphenyl carbamate based Legumain inhibitors.
Bioorganic & medicinal chemistry letters (2014-04-30)
Catherine Higgins, Samira Bouazzaoui, Kishore Gaddale, Zenobia D'Costa, Amy Templeman, Martin O'Rourke, Andrew Young, Christopher Scott, Tim Harrison, Paul Mullan, Rich Williams
ZUSAMMENFASSUNG
This Letter describes the further development and SAR exploration of a novel series of Legumain inhibitors. Based upon a previously identified Legumain inhibitor from our group, we explored the SAR of the carbamate phenyl ring system to probe the P3 pocket of the enzyme. This led to the identification of a sub-nanomolar inhibitor of Legumain.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Discovery® Cyano HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® Cyano Supelguard Kartusche, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® Cyano HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4 mm