Skip to Content
Merck

90605

1,5,7-Triazabicyclo[4.4.0]dec-5-ene

purum, ≥98.0% (T)

Synonym(s):

1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine, Hhpp, TBD

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

Empirical Formula (Hill Notation):
C7H13N3
CAS Number:
Molecular Weight:
139.20
Beilstein:
3242
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

purum

Assay

≥98.0% (T)

mp

125-130 °C (lit.)
125-132 °C

solubility

toluene: 1 g/15 mL

SMILES string

C1CNC2=NCCCN2C1

InChI

1S/C7H13N3/c1-3-8-7-9-4-2-6-10(7)5-1/h1-6H2,(H,8,9)

InChI key

FVKFHMNJTHKMRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Guanidine base which is about 100 times more basic than tetramethylguanidine; Powerful base for tautomerizing pyrrocorphins and for other reactions

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U. Kampfen et al.
Helvetica Chimica Acta, 72, 185-185 (1989)
K. Hilpert et al.
Journal of the Chemical Society. Chemical Communications, 1401-1401 (1983)
R. Schwesinger
Chimia, 39, 269-269 (1985)
Jinfeng Wang et al.
PloS one, 10(2), e0118086-e0118086 (2015-02-24)
MicroRNA-214 (miR-214) has been reported to be dysregulated in human bladder cancer tissues. We aimed to investigate the clinical correlation, biological significance and molecular network of miR-214 in bladder cancer. Our results showed miR-214 was down-regulated in bladder cancer tissues
Shouru Zhang et al.
Medical oncology (Northwood, London, England), 32(1), 414-414 (2014-12-05)
The mammalian peroxiredoxin 2 (Prdx2) is a member of thiol-dependent antioxidant proteins and plays an important role in the progression of colorectal cancer (CRC). The aim of this study was to confirm the role of Prdx2 in formation of VM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service