Skip to Content
Merck

Skip To

EHU028811

MISSION® esiRNA

targeting human EEF1AKMT1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
€ 219,00
50 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
€ 394,00

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324
eCl@ss:
32119207

€ 219,00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Powered by Eupheria Biotech

Quality Segment

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCAACTGAGCCAGTTTTGGTATAGTCAGGAAACTGCTCTGCAGCTGGCACAGGAGGCAATTGCAGCTGTAGGAGAAGGTGGCAGAATCGCATGTGTGAGTGCCCCTAGTGTTTACCAGAAACTCAGAGAGCTGTGCAGAGAAAACTTTTCGATATACATCTTTGAATATGACAAAAGATTTGCCATGTATGGAGAGGAGTTTATTTTCTATGATTACAATAATCCATTGGACTTACCCGAAAGAATTGCTGCACATAGTTTTGACATCGTAATAGCAGATCCTCCCTATCTTTCGGAGGAATGTCTCAGAAAAACATCGGAAACCGTCAAGTACCTGACGCGGGGCAAGATTCTGCTGTGCACAGGTGCCATCATGGAAGAACAGGCAGCAGAACTCCTTGGAGTGAAGATGTGCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EMU015871HPA040417HPA042543
description

targeting human EEF1AKMT1

description

targeting mouse N6amt2

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

esiRNA cDNA target sequence

GCAACTGAGCCAGTTTTGGTATAGTCAGGAAACTGCTCTGCAGCTGGCACAGGAGGCAATTGCAGCTGTAGGAGAAGGTGGCAGAATCGCATGTGTGAGTGCCCCTAGTGTTTACCAGAAACTCAGAGAGCTGTGCAGAGAAAACTTTTCGATATACATCTTTGAATATGACAAAAGATTTGCCATGTATGGAGAGGAGTTTATTTTCTATGATTACAATAATCCATTGGACTTACCCGAAAGAATTGCTGCACATAGTTTTGACATCGTAATAGCAGATCCTCCCTATCTTTCGGAGGAATGTCTCAGAAAAACATCGGAAACCGTCAAGTACCTGACGCGGGGCAAGATTCTGCTGTGCACAGGTGCCATCATGGAAGAACAGGCAGCAGAACTCCTTGGAGTGAAGATGTGCA

esiRNA cDNA target sequence

AGGACACTGCTTTGCGACTTGCACGGGAAGCGATCGATGCTGCTGGAGAAGGAGGCAGAATCGCGTGTGTGAGCGCGCCAAGCGTGTACCAGAAGCTCCGGGAGCTCTGTAGGGAAGACAGCTCTGTCTACATCTTTGAATACGACAGAAGATTCGCCATATATGGGGATGAATTTATCTTCTATGATTACAACCATCCACTGGAGTTGCCTGAGAGAATCGCTGCGCATAGCTTTGACCTTGTGGTTGCAGACCCTCCTTACCTGTCTGAGGAATGTCTCAGGAAGACATCTGAAACGATCCAGTTTCTGACTCGGGGCAAGATTCTGCTGTGCACAGGAGCCATCATGGAAGAGCAGGCAGCTCAGCTCCTGGGTGTGAAGATGTGCAAGTTTATTCCGGAACACTCCCGAAACTTGGCCAATGAGTT

esiRNA cDNA target sequence

-

esiRNA cDNA target sequence

-

Ensembl | human accession no.

ENSG00000150456

Ensembl | human accession no.

-

Ensembl | human accession no.

-

Ensembl | human accession no.

-

product line

MISSION®

product line

MISSION®

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

Gene Information

human ... EEF1AKMT1(221143), N6AMT2(221143)

Gene Information

mouse ... N6AMT2(68043), N6amt2(68043)

Gene Information

human ... N6AMT2(221143)

Gene Information

human ... N6AMT2(221143)

NCBI accession no.

NM_174928

NCBI accession no.

NM_026526

NCBI accession no.

-

NCBI accession no.

-

form

lyophilized powder

form

lyophilized powder

form

-

form

buffered aqueous glycerol solution


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters