Skip to Content
Merck

Skip To

EMU020081

MISSION® esiRNA

targeting mouse Thoc6

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
€ 219,00
50 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
€ 394,00

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324
eCl@ss:
32119207

€ 219,00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Powered by Eupheria Biotech

Quality Segment

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCAACAATTACGGGCAGATTGCCATCTTCAGTTTGTCTGCTGCCTTAAGTTCAGAGGCCAAAGAAGAAAGCAAAAAACCGGTGGTGGTCTTCCACGCCCATGACGGGCCTGTCTACAGCATGGTCTCCACAGATCGACACCTACTCAGTGCTGGCGATGGAGAGGTCAAGGGTTGGCTTTGGGCAGAAATTCTCAAGAAGGGCTGTAAGGAACTGTGGCGTCGCCAGCCTCCATACAGGACCAGTCTGGAAGTACCCGAAATCAATGCCTTGCTGCTTGTCCCCAAGGAGAATTCGCTCATCCTGGCTGGGGGAGATTGTCAGCTGCACAGTATGGATCTGGAAACTGGGGCCTTCACACGGGCCCTCCGGGGCCACACAGACTACATCCATTGCCTGGCACTGAGGGAGCGCAGCCCTGAGGTATTGTCTGGCGGTGAG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EHU094551HPA052953
description

targeting mouse Thoc6

description

targeting human THOC6

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

product line

MISSION®

product line

MISSION®

product line

Prestige Antibodies® Powered by Atlas Antibodies

esiRNA cDNA target sequence

GCAACAATTACGGGCAGATTGCCATCTTCAGTTTGTCTGCTGCCTTAAGTTCAGAGGCCAAAGAAGAAAGCAAAAAACCGGTGGTGGTCTTCCACGCCCATGACGGGCCTGTCTACAGCATGGTCTCCACAGATCGACACCTACTCAGTGCTGGCGATGGAGAGGTCAAGGGTTGGCTTTGGGCAGAAATTCTCAAGAAGGGCTGTAAGGAACTGTGGCGTCGCCAGCCTCCATACAGGACCAGTCTGGAAGTACCCGAAATCAATGCCTTGCTGCTTGTCCCCAAGGAGAATTCGCTCATCCTGGCTGGGGGAGATTGTCAGCTGCACAGTATGGATCTGGAAACTGGGGCCTTCACACGGGCCCTCCGGGGCCACACAGACTACATCCATTGCCTGGCACTGAGGGAGCGCAGCCCTGAGGTATTGTCTGGCGGTGAG

esiRNA cDNA target sequence

GGAGCACTCTGGGACTTGTAGTTCTGGAGATGGAGCGAGCTGTGCCGCTCGCGGTGCCTCTGGGTCAGACAGAGGTGTTCCAGGCCTTGCAGCGGCTCCATATGACCATCTTCTCCCAGAGCGTCTCACCATGTGGGAAGTTTCTGGCGGCTGGCAACAATTACGGGCAGATTGCCATCTTCAGCTTGTCCTCTGCTTTGAGCTCAGAAGCCAAAGAGGAAAGTAAGAAGCCGGTGGTGACTTTCCAAGCCCATGATGGGCCCGTCTATAGCATGGTTTCCACCGATCGACATCTGCTTAGTGCTGGGGATGGGGAGGTGAAGGCCTGGCTTTGGGCGGAGATGCTCAAGAAGGGCTGTAAGGAGCTGTGGCGTCGTCAGCCTCCATACAGGACCAGCCTGGAAGTGCCTGAGATCAACGCT

esiRNA cDNA target sequence

-

Ensembl | mouse accession no.

ENSMUSG00000041319

Ensembl | mouse accession no.

-

Ensembl | mouse accession no.

-

Gene Information

mouse ... THOC6(386612), Thoc6(386612)

Gene Information

human ... THOC6(79228)

Gene Information

human ... THOC6(79228)

form

lyophilized powder

form

lyophilized powder

form

buffered aqueous glycerol solution

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters