Skip to Content
Merck

EMU024941

MISSION® esiRNA

targeting mouse Nuf2

Sign In to View Organizational & Contract Pricing.

Select a Size

20 μG

€ 212,00

50 μG

€ 382,00

€ 212,00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


About This Item

NACRES:
NA.51
UNSPSC Code:
41105324

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

MISSION® esiRNA, targeting mouse Nuf2

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCAGGAGGACCTTGTTGAGCCTTTCCTGCGTCTACAGAGGTCTGTGGTTTCTTGAAACACAGGAAGCAACAATACTCAGGGAGGAGACTCCCATAATGGAAACTTTGTCATTCCCCAGATACAATGTAGCTGAGCTTGTGGTTCATATTCGCAATAAATTATTAACAGGAGCAGATGGCAAAAACCTCTCGAAGAGCGATTTTCTTCCAAATCCGAAGTCTGATGTCTTGTACATGATTTACATGAAAGCCTTACAGTTAGTGTATGGAGTCCGGCTGGAGCACTTCTACATGATGCCGATGAACATAGAAGTCACATATCCACATTTAATGGAGGGCTTCTTACCAGTCAGAAGTTTGTTCTTTTACATGGATTCCTTTATGCCCATTTGCCGGGTGAATGACTTTGAGATTGTCGACATTCTTAATCCAAGAACAAACCGGACAAGTCGT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Quality Level

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EMU091561EMU003431EMU046461
Gene Information

mouse ... NUF2(66977), Nuf2(66977)

Gene Information

mouse ... NUTF2(68051), Nutf2(68051)

Gene Information

mouse ... CNTNAP2(66797), Cntnap2(66797)

Gene Information

mouse ... NDUFC1(66377), Ndufc1(66377)

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

Ensembl | mouse accession no.

ENSMUSG00000026683

Ensembl | mouse accession no.

ENSMUSG00000008450

Ensembl | mouse accession no.

ENSMUSG00000039419

Ensembl | mouse accession no.

ENSMUSG00000037152

esiRNA cDNA target sequence

TCCAGGAGGACCTTGTTGAGCCTTTCCTGCGTCTACAGAGGTCTGTGGTTTCTTGAAACACAGGAAGCAACAATACTCAGGGAGGAGACTCCCATAATGGAAACTTTGTCATTCCCCAGATACAATGTAGCTGAGCTTGTGGTTCATATTCGCAATAAATTATTAACAGGAGCAGATGGCAAAAACCTCTCGAAGAGCGATTTTCTTCCAAATCCGAAGTCTGATGTCTTGTACATGATTTACATGAAAGCCTTACAGTTAGTGTATGGAGTCCGGCTGGAGCACTTCTACATGATGCCGATGAACATAGAAGTCACATATCCACATTTAATGGAGGGCTTCTTACCAGTCAGAAGTTTGTTCTTTTACATGGATTCCTTTATGCCCATTTGCCGGGTGAATGACTTTGAGATTGTCGACATTCTTAATCCAAGAACAAACCGGACAAGTCGT

esiRNA cDNA target sequence

AGCCTTCCGTTCCAGAAAATTCAGCATAGCATCACGGCGCAGGACCACCAGCCTACACCAGATAGCTGCATCATCAGCATGGTTGTGGGCCAGCTCAAGGCTGATGAAGATCCCATCATGGGTTTCCACCAGATGTTTCTATTAAAGAACATCAACGATGCTTGGGTTTGCACCAATGACATGTTCAGGCTTGCCCTGCACAACTTCGGCTGACCTCCTCCTGGCCACAGTCACGCTGTTTCCTCCTCCCTCTTCCCAATCCTAGCTCACTCCTCCAGATGCTCCAAATATCATACACAAACGAGCAGGGCCGAGGCGGGAGCGGGTGCAGGGCGCTTCTGTCACCACGGTGTTGTGCATGATGTTTGGATGCTAGACTAGTCGCATCTGACAGGAAGTTTGTGTTGTACCAGCGCAT

esiRNA cDNA target sequence

CACACCTGAGCAAGCTGAAAGAAAGAGAAACGGGAAAAATCGACCAAGAAATTCACAAATACAACACCCCAGGCTTCACAGGCTGCCTGTCCAGAGTCCAGTTCAACCACATCGCCCCTCTCAAGGCAGCCTTGAGACAGACAAACGCCTCAGCCCACGTGCACATTCAGGGTGAGCTGGTGGAATCTAACTGCGGGGCCTCCCCACTGACGCTTTCCCCCATGTCTTCAGCCACTGACCCTTGGCACCTAGATCACTTGGATTCAGCCAGTGCTGATTTCCCATATAACCCAGGACAAGGCCAAGCTATAAGAAATGGAGTCAACAGAAACTCAGCTATCATTGGAGGGGTCATCGCTGTGGTGATTTTCACCATCCTCTGCACCCTGGTCTTCCTCATCCGGTA

esiRNA cDNA target sequence

AGCCTGCGGAACAAGATGGCGCCGTCCGTAGTGCTGCGCTCGTTTTCGCGGCTGCTGGCCCCCGCCCGGCTGCCGAGCTGCTCTTCAACACGGTCGAAGTTCTATGTCCGGGAGCCAGTCAATGCCAAACCTAACTGGTTGGCAGTTGGACTGTCCGTGGGCGCCTCGGTTTTCATGTGGATTTATCTCATCCAAACACACAATGAAGATGTTTTGGAGTATAAAAGAAGAAATGGATTGGAATAAACTTTGGAAATAATACAGTCAGCTTCATGTGATGATCACAGTCCTTCCTCTGGATTCACCCTCAGTGACGGATGTGAGAAAAGAGTTACATGCGAATATGAGTCAGTCAACACGTTTTGCATGGTTAAGTTAAAGAAATAAAAATACTCGCGCTCCTC

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service