Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | € 219,00 |
50 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | € 394,00 |
About This Item
€ 219,00
description
Powered by Eupheria Biotech
Quality Segment
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
AGCGATTTATGCACCTCAGAATGTCTACTGTATCCACAGTGATGAAAATGCCCCAAAGAAGTTTAAGAGTGCCATGCAAACCTTCGTGGACTGTTTTGGAAACATCTTTCTTTCATCAAAGACACAGAAGGTGGCTCATGACAACCTCAGGAGGCTGCAGGCAGAGATTGACTGCATGAGGGACCTGGTCCACTCCCCATTTCAATGGCACTATGTCATGAATCTCTGTGGACAGGAATTCCCTATCAAAACCAACAAAGAAATCATCTATGACATCAGAACCAGGTGGAAGGGTAAAAACATCACGCCTGGGGTGACCCCACCCGCAAACTCCAAACCCAAGACAGGCCAAGGCCCTCCAAAGCCCAGTCCCGATGAAAACAGCTACACAGCTCCAAATACAATTTTCAAACAAAGCCCACCCCACAATCTAACAATTTCCTCTGGAAGTGCTCACTATGCACTTACGAGGAAGTTTGTAGAGTTCGTCCTAACTGATCCCCGTGCAAA
Ensembl | mouse accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
mouse ... GCNT7(654821), Gcnt7(654821)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description targeting mouse Gcnt7 | description targeting human GCNT7 | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution |
| product line MISSION® | product line MISSION® | product line Prestige Antibodies® Powered by Atlas Antibodies | product line Prestige Antibodies® Powered by Atlas Antibodies |
| esiRNA cDNA target sequence AGCGATTTATGCACCTCAGAATGTCTACTGTATCCACAGTGATGAAAATGCCCCAAAGAAGTTTAAGAGTGCCATGCAAACCTTCGTGGACTGTTTTGGAAACATCTTTCTTTCATCAAAGACACAGAAGGTGGCTCATGACAACCTCAGGAGGCTGCAGGCAGAGATTGACTGCATGAGGGACCTGGTCCACTCCCCATTTCAATGGCACTATGTCATGAATCTCTGTGGACAGGAATTCCCTATCAAAACCAACAAAGAAATCATCTATGACATCAGAACCAGGTGGAAGGGTAAAAACATCACGCCTGGGGTGACCCCACCCGCAAACTCCAAACCCAAGACAGGCCAAGGCCCTCCAAAGCCCAGTCCCGATGAAAACAGCTACACAGCTCCAAATACAATTTTCAAACAAAGCCCACCCCACAATCTAACAATTTCCTCTGGAAGTGCTCACTATGCACTTACGAGGAAGTTTGTAGAGTTCGTCCTAACTGATCCCCGTGCAAA | esiRNA cDNA target sequence GGCGATTTCTCTTTGGCATATATTATAACTATTCATAAGGAGCTGGCCATGTTTGTGCAGCTTCTCAGAGCTATTTATGTACCTCAAAATGTTTATTGTATTCATGTTGATGAAAAGGCCCCAATGAAGTATAAGACTGCTGTGCAAACCTTGGTTAACTGTTTTGAAAACGTTTTTATTTCCTCCAAGACAGAGAAGGTGGCTTATGCTGGCTTTACAAGACTACAGGCAGATATTAATTGTATGAAAGTTCTAGTGCATTCTAAATTTCAATGGAACTATGTCATCAATCTTTGTGGACAGGATTTTCCCATCAAAACCAACAGAGAAATCATACACTACATCAGAAGCAAATGGAGTGATAAAAATATTACTCCTGGAGTAATCCAACCATTGCACATTAAATCCAAGACAAGTCAAAGTCATCTCGAATTCGTTCCTAAAGGAAGTATCTATGCACCTCCAAATAACAGATTCAAAGACAAACCACCCCAT | esiRNA cDNA target sequence - | esiRNA cDNA target sequence - |
| Ensembl | mouse accession no. | Ensembl | mouse accession no. - | Ensembl | mouse accession no. - | Ensembl | mouse accession no. - |
| Gene Information mouse ... GCNT7(654821), Gcnt7(654821) | Gene Information human ... GCNT7(140687) | Gene Information human ... GCNT7(140687) | Gene Information human ... GCNT7(140687) |
| NCBI accession no. | NCBI accession no. - | NCBI accession no. - | NCBI accession no. - |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


