Skip to Content
Merck

Skip To

O0876

Oleoyl chloride

≥95% (GC)

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COCl
CAS Number:
Molecular Weight:
300.91
EC Number:
204-005-0
UNSPSC Code:
12352211
PubChem Substance ID:
Beilstein/REAXYS Number:
1211748
MDL number:


assay

≥95% (GC)

refractive index

n20/D 1.463 (lit.)

bp

193 °C/4 mmHg (lit.)

density

0.91 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(Cl)=O

InChI

1S/C18H33ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3/b10-9-

InChI key

MLQBTMWHIOYKKC-KTKRTIGZSA-N

Packaging

Sealed ampule.


Still not finding the right product?

Explore all of our products under Oleoyl chloride


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

339.8 °F - closed cup

flash_point_c

171 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Noemi Jiménez-Rojo et al.
Current biology : CB, 30(19), 3775-3787 (2020-08-29)
Sphingolipids play important roles in physiology and cell biology, but a systematic examination of their functions is lacking. We performed a genome-wide CRISPRi screen in sphingolipid-depleted human cells and identified hypersensitive mutants in genes of membrane trafficking and lipid biosynthesis
Kui Yang et al.
Analytical and bioanalytical chemistry, 407(17), 5199-5210 (2015-03-31)
Diglycerides play a central role in lipid metabolism and signaling in mammalian cells. Although diacylglycerol molecular species comprise the majority of cellular diglycerides that are commonly measured using a variety of approaches, identification of extremely low abundance vinyl ether diglycerides



Questions

Reviews

No rating value

Active Filters