Skip to Content
Merck

O7128

Octanal

Synonym(s):

Aldehyde C8, Caprylic aldehyde, Octyl aldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
CH3(CH2)6CHO
CAS Number:
Molecular Weight:
128.21
EC Number:
204-683-8
UNSPSC Code:
12352200
MDL number:
Beilstein/REAXYS Number:
1744086
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C8H16O/c1-2-3-4-5-6-7-8-9/h8H,2-7H2,1H3

InChI key

NUJGJRNETVAIRJ-UHFFFAOYSA-N

SMILES string

[H]C(=O)CCCCCCC

vapor pressure

2 mmHg ( 20 °C), 850 mmHg ( 25 °C)

refractive index

n20/D 1.421 (lit.)

bp

171 °C (lit.)

mp

12-15 °C (lit.)

density

0.82 g/mL at 25 °C (lit.), 0.82 g/mL at 25 °C (lit.)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service