Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
Empirical Formula (Hill Notation):
C36H69NO3
CAS Number:
Molecular Weight:
563.94
UNSPSC Code:
12352200
MDL number:
SMILES string
CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)NCCCCCCC\C=C\CCCCCCCC
storage temp.
−20°C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service