Merck
  • Dietary Lipids Induce Ferroptosis in Caenorhabditiselegans and Human Cancer Cells.
Dietary Lipids Induce Ferroptosis in Caenorhabditiselegans and Human Cancer Cells.
Developmental cell (2020-07-12)
Marcos A Perez, Leslie Magtanong, Scott J Dixon, Jennifer L Watts
ABSTRACT
Dietary lipids impact development, homeostasis, and disease, but links between specific dietary fats and cell fates are poorly understood. Ferroptosis is an iron-dependent form of nonapoptotic cell death associated with oxidized polyunsaturated phospholipids. Here, we show that dietary ingestion of the polyunsaturated fatty acid (PUFA) dihomogamma-linolenic acid (DGLA; 20:3n-6) can trigger germ-cell ferroptosis and sterility in the nematode Caenorhabditis elegans. Exogenous DGLA is also sufficient to induce ferroptosis in human cells, pinpointing this omega-6 PUFA as a conserved metabolic instigator of this lethal process. In both C. elegans and human cancer cells, ether-lipid synthesis protects against ferroptosis. These results establish C. elegans as a powerful animal model to study the induction and modulation of ferroptosis by dietary fats and indicate that endogenous ether lipids act to prevent this nonapoptotic cell fate.
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
2,2′-Bipyridyl, ReagentPlus®, ≥99%
Sigma-Aldrich
Ferrostatin-1, ≥95% (HPLC)
Sigma-Aldrich
TERGITOL solution, Type NP-40, 70% in H2O