Merck
Clinical benefits of low serum digoxin concentrations in heart failure.
Journal of the American College of Cardiology (2002-03-19)
Kirkwood F Adams, Mihai Gheorghiade, Barry F Uretsky, J Herbert Patterson, Todd A Schwartz, James B Young
ABSTRACT
We sought to determine whether there was a relationship between serum digoxin concentration (SDC), including SDCs typically regarded as low, and clinical efficacy related to digoxin in patients with symptomatic left ventricular dysfunction. Digitalis glycosides have been used for 200 years in the treatment of heart failure (HF), but the SDC required for optimal clinical efficacy and acceptable toxicity remains controversial. This relationship was investigated by utilizing data from two randomized, double-blinded, placebo-controlled, digoxin-withdrawal trials: the Prospective Randomized study Of Ventricular failure and Efficacy of Digoxin (PROVED) and the Randomized Assessment of Digoxin on Inhibitors of Angiotensin-Converting Enzyme (RADIANCE). Major end points were worsening HF, change in left ventricular ejection fraction and treadmill time after randomization. The primary analysis investigated the relationship between SDC at randomization and these end points. A secondary categorical analysis compared these end points in patients who discontinued digoxin versus patients who continued digoxin and had low (0.5 to 0.9 ng/ml), moderate (0.9 to 1.2 ng/ml) or high (>1.2 ng/ml) SDCs at randomization. Multiple regression analysis failed to find a relationship between randomization SDC, considered as a continuous variable, and any study end point (all p > 0.236). Multivariable Cox analysis found that the risk of worsening HF was significantly less (all p < 0.02) for patients in any category of SDC who continued digoxin, as compared with patients withdrawn from digoxin. Specifically, patients in the low SDC category were significantly less likely than placebo patients to experience worsening HF during follow-up (p = 0.018). The beneficial effects of digoxin on common clinical end points in patients with HF were similar, regardless of SDC.
Sigma-Aldrich
Taurine, ≥98%, FG
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
SAFC
Taurine
SAFC
L-Isoleucine
SAFC
L-Methionine
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Methionine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material