Merck
Short- and long-term effects of acetaldehyde on plasma.
Alcohol (Fayetteville, N.Y.) (2002-04-18)
Arthur S Brecher, Mohammed T Adamu
ABSTRACT
The effect of low concentrations of acetaldehyde on activated partial thromboplastin time (APTT) and prothrombin time (PT) of Accuclot coagulation plasmas was monitored over a prolonged time to mimic effects observed in alcoholism. A prolongation of the APTT from 31.9 +/- 0.7 s to 32.6 +/- 0.9 s (n = 8; P =.007) was observed after a 30-min preincubation time with 140 microM acetaldehyde. However, a minimum of 3.6 mM acetaldehyde was required to extend the APTT from 36.6 +/- 1.0 s to 41.2 +/- 0.8 s (P =.001) over an 18-h exposure time. Plasma acetaldehyde levels as low as 2.24 mM caused elevation of PTs from 12.5 +/- 0.5 s to 14.4 +/- 0.2 s (P =.005) after a 24-h preincubation time. These findings seem to indicate that short-term contact of acetaldehyde with plasma, probably yielding reversible interactions, may interfere with APTTs to a greater extent than long-term contact, which would presumably yield stable, irreversible interactions. In comparing the effects of 8.94, 17.9, 89.4, and 447 mM acetaldehyde on the PTs of Level I, II, and III plasma, the PTs were most increasingly prolonged in Level III plasma and least prolonged in Level I plasma at each acetaldehyde concentration, although the plasmas have comparable protein concentrations. These findings seem to indicate that coagulation factors are sensitive to inactivation by acetaldehyde.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Supelco
2-Butanone, analytical standard
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
5α-Pregnane-3,20-dione (allo), progesterone metabolite
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Progesterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®