Merck
  • In vivo detection of brain glycine with echo-time-averaged (1)H magnetic resonance spectroscopy at 4.0 T.
In vivo detection of brain glycine with echo-time-averaged (1)H magnetic resonance spectroscopy at 4.0 T.
Magnetic resonance in medicine (2006-02-03)
Andrew P Prescot, Blaise de B Frederick, Liqun Wang, John Brown, J Eric Jensen, Marc J Kaufman, Perry F Renshaw
ABSTRACT
A single-voxel proton magnetic resonance spectroscopy ((1)H-MRS) method is described that enables the in vivo measurement of endogenous brain glycine (Gly) levels in human subjects. At 4.0 T, TE-averaging (1)H-MRS dramatically attenuates the overlapping myo-inositol (mI) resonances at 3.52 ppm, permitting a more reliable measure of the Gly singlet peak. This methodology initially is described and tested in phantoms. The phantom data infers that the 3.55-ppm peak predominantly is Gly with a smaller contribution from mI. The composite resonance thus is differentiated from pure Gly and mI and is labeled Gly*. The mI contribution was calculated as <2% of the total Gly* signal for a 1:1 mI/Gly mixture. The technique subsequently was used to acquire TE-averaged (1)H-MRS data from the occipital cortex of healthy control subjects. The resultant spectra closely resembled experimental phantom data. LC-model analysis provided a means for quantifying TE-averaged (1)H-MRS spectra and a mean test-retest variability measure of 15% was established for brain Gly* levels in studies of six healthy subjects.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
SAFC
Glycine
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine 1 M solution