Merck
Understanding the human salivary metabolome.
NMR in biomedicine (2009-03-05)
Ienaka Takeda, Cynthia Stretch, Pamela Barnaby, Kriti Bhatnager, Kathryn Rankin, Hao Fu, Aalim Weljie, Naresh Jha, Carolyn Slupsky
ABSTRACT
Saliva is a readily accessible biofluid that is important for the overall health, aiding in the chewing, swallowing, and tasting of food as well as the regulation mouth flora. As a first step to determining and understanding the human saliva metabolome, we have measured salivary metabolite concentrations under a variety of conditions in a healthy population with reasonably good oral hygiene. Using (1)H NMR spectroscopy, metabolite concentrations were measured in resting (basal) and stimulated saliva from the same subject and compared in a cohort of healthy male non-smoking subjects (n = 62). Almost all metabolites were higher in the unstimulated saliva when compared to the stimulated saliva. Comparison of the salivary metabolite profile of male smokers and non-smokers (n = 46) revealed citrate, lactate, pyruvate, and sucrose to be higher and formate to be lower in concentration in smokers compared with non-smokers (p < 0.05). Gender differences were also investigated (n = 40), and acetate, formate, glycine, lactate, methanol, propionate, propylene glycol, pyruvate, succinate, and taurine were significantly higher in concentration in male saliva compared to female saliva (p < 0.05). These results show that differences between male and female, stimulated and unstimulated, as well as smoking status may be observed in the salivary metabolome.
Product Description
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Dimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Supelco
Acetone, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)