Merck
  • Sivelestat attenuates lung injury in surgery for congenital heart disease with pulmonary hypertension.
Sivelestat attenuates lung injury in surgery for congenital heart disease with pulmonary hypertension.
The Annals of thoracic surgery (2013-10-01)
Norikazu Nomura, Miki Asano, Takayuki Saito, Takuya Nakayama, Akira Mishima
ABSTRACT
Pulmonary hypertension associated with congenital heart disease increases the risk of surgery using cardiopulmonary bypass. Sivelestat is a neutrophil elastase inhibitor thought to have a prophylactic effect against lung injury after surgery using bypass. We elucidated that Sivelestat had the protective effect on lung in patients with congenital heart disease and pulmonary hypertension who underwent surgery using bypass. This study was a controlled prospective randomized trial and enrolled 13 neonates or infants with ventricular septal defect and pulmonary hypertension. The patients were assigned to either sivelestat with the dose of 0.2 mg/kg per hour (sivelestat group, n = 7) or saline (placebo group, n = 6) from the start of bypass until 6 hours after bypass. Proinflammatory cytokines and adhesion molecules on leukocytes were measured at 10 time points during the above period. Pulmonary function was assessed perioperatively. Compared with the placebo group, the sivelestat group had significantly lower values of alveolar-arterial oxygen tension gradient at 24 hours (p = 0.038) and at 48 hours (p = 0.028) after bypass, and significantly better balance of hydration at 48 hours after bypass (p = 0.012). The sivelestat group also showed significantly lower plasma levels of interleukin-8 immediately after bypass (p = 0.041) and interleukin-10 at 15 minutes after removal of the aortic cross-clamp (p = 0.048), and immediately after bypass (p = 0.037). Administration of sivelestat during bypass prevented pulmonary damage and activities of proinflammatory cytokines at the cardiac operation in neonates or infants. Our results show that sivelestat may be considered to protect pulmonary function against the injury by bypass.
Product Description
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
Glycine, 99%, FCC
SAFC
Glycine
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Sivelestat sodium salt hydrate, ≥98% (HPLC), solid
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)
Sigma-Aldrich
Glycine 1 M solution