Merck
  • T-2 toxin regulates steroid hormone secretion of rat ovarian granulosa cells through cAMP-PKA pathway.
T-2 toxin regulates steroid hormone secretion of rat ovarian granulosa cells through cAMP-PKA pathway.
Toxicology letters (2014-12-30)
Jing Wu, Di Tu, Li-Yun Yuan, Jin-e Yi, Yanan Tian
ABSTRACT
T-2 toxin is a secondary metabolite produced by Fusarium genus and is a common contaminant in food and feedstuffs of cereal origin. In porcine granulosa cells(GC), T-2 toxin has been shown to inhibit the steroidogenesis; however, the mechanism has not been well understood. Gonadotropin-stimulated steroidogenesis is regulated by the cAMP-PKA pathway. In this study, we investigated potential mechanisms for T-2 toxin-induced reproductive toxicity focusing on the critical steps of the cAMP-PKA pathway affected by T-2 toxin. We first analyzed the effects of T-2 toxin on progesterone and estrogen production in rat granulosa cells. For this purpose the granulosa cells were cultured for 48 h in 10% fetal bovine serum-containing medium followed by 24h in serum-free medium containing FSH (10 ng/ml) and androstenedione (3 ng/ml), both are required for normal steroidogenesis. Treatment of these cells with T-2 toxin dose-dependently inhibited the growth of cells and the steroid hormone production. Cellular cyclic AMP levels were dose-dependently inhibited by T-2 toxin (0, 1, 10 and 100 nM, 24 h). Furthermore, we found that although the induction of progesterone by 8-Br-cAMP (a FSH mimetic) and 22R-HC (substrate for progesterone) could both be inhibited by T-2 toxin treatment, the T-2-imposed inhibitory effects could be reversed by increasing doses of 22R-HC, while increasing 8-Br-cAMP had no effects, suggesting that T2 toxin targeted at distinct mechanisms. cAMP-stimulated steroidogenic acute regulatory protein (StAR) is a rate limiting protein in progesterone synthesis. Exposure to T2 toxin caused significant suppression of StAR expression as determined by Western blotting and semi-quantitative RT-PCR suggesting StAR is a sensitive target for T-2 toxin. Taken together, our results strongly suggest that T2 toxin inhibits steroidogenesis by suppressing cAMP-PKA pathway and StAR is a target for T-2-toxin. The antisteroidogenesis effects were observable at low T-2 dose (1 ng/ml) suggesting T-2 toxin has an endocrine disruptive effect.
Product Description
Supelco
T2-Toxin solution, 100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
8-Bromoadenosine 3′,5′-cyclic monophosphate, ≥97% (HPLC)
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
T-2 Toxin, from Fusarium sp., powder, ≥98% (HPLC)
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Supelco
Progesterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
β-Estradiol, ≥98%
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
Estradiol, meets USP testing specifications
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, 99%, FCC
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Glycine
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)