Merck
  • Citrulline induces fatty acid release selectively in visceral adipose tissue from old rats.
Citrulline induces fatty acid release selectively in visceral adipose tissue from old rats.
Molecular nutrition & food research (2014-06-11)
Nolwenn Joffin, Anne-Marie Jaubert, Sylvie Durant, Jean Bastin, Jean-Pascal De Bandt, Luc Cynober, Christophe Moinard, Claude Forest, Philippe Noirez
ABSTRACT
During aging, increased visceral adipose tissue (AT) mass may result in impaired metabolic status. A citrulline (CIT)-supplemented diet reduces AT mass in old rats. We hypothesized that CIT could directly affect fatty acid (FA) metabolism in retroperitoneal AT. A 24-h exposure of AT explants from old (25 months) rats to 2.5 mM CIT induced a 50% rise in glycerol and FA release, which was not observed in explants from young (2 months) animals. The phosphorylated form of hormone-sensitive lipase, a key lipolytic enzyme, was 1.5-fold higher in CIT-treated explants from old and young rats, whereas glyceroneogenesis, that provides glycerol-3P requested for FA re-esterification, and its key enzyme phosphoenolpyruvate carboxykinase, were down-regulated 40-70%. Specifically in young rats, beta-oxidation capacity and gene expressions of carnitine palmitoyl transferase 1-b and very long chain acyl-CoA dehydrogenase were strongly up-regulated by CIT. In contrast, in old rats, while glyceroneogenesis was lower, beta-oxidation was not affected, enabling increased FA release. Hence, in visceral AT, CIT exerts a specific induction of the beta-oxidation capacity in young rats and a selective stimulation of FA release in old rats, therefore providing a direct mechanism of CIT action to reduce AT mass.
Product Description
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Supelco
Palmitic acid, analytical standard