Merck
  • Altered VEGF-stimulated Ca2+ signaling in part underlies pregnancy-adapted eNOS activity in UAEC.
Altered VEGF-stimulated Ca2+ signaling in part underlies pregnancy-adapted eNOS activity in UAEC.
The Journal of endocrinology (2014-07-27)
Derek S Boeldt, Mary A Grummer, Ronald R Magness, Ian M Bird
ABSTRACT
In pregnancy, the uterine vasculature undergoes dramatic vasodilatory adaptations. Previously, vascular endothelial growth factor (VEGF) has been shown to stimulate endothelial nitric oxide synthase (eNOS) in uterine artery endothelial cells (UAECs) derived from pregnant ewes to a greater extent than those from non-pregnant ewes in a manner not fully explained by changes in the phosphorylation of eNOS. In this study, we used Fura-2 Ca(2+) imaging and arginine-to-citrulline conversion eNOS activity assays to assess the importance of VEGF-stimulated Ca(2+) responses in pregnancy-related changes in NO production in UAEC. In this study, we show that pregnancy-induced changes in VEGF-stimulated Ca(2+) responses could account in part for the greater capacity of VEGF to stimulate eNOS in UAECs from pregnant versus non-pregnant animals. VEGF-stimulated Ca(2+) responses in UAECs from pregnant and non-pregnant animals were mediated through VEGF receptor 2 and were detected in roughly 15% of all cells. There were no pregnancy-specific differences in area under the curve or peak height. UAECs from pregnant animals were more consistent in the time to response initiation, had a larger component of extracellular Ca(2+) entry, and were more sensitive to a submaximal dose of VEGF. In UAECs from pregnant and non-pregnant animals Ca(2+) responses and eNOS activation were sensitive to the phospholipase C/inositol 1,4,5-trisphosphate pathway inhibitors 2-aminoethoxydiphenylborane and U73122. Thus, changes in VEGF-stimulated [Ca(2+)]i are necessary for eNOS activation in UAECs, and pregnancy-induced changes in Ca(2+) responses could also in part explain the pregnancy-specific adaptive increase in eNOS activity in UAECs.
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis