Merck
  • Phosphate modulates receptor sulfotyrosine recognition by the chemokine monocyte chemoattractant protein-1 (MCP-1/CCL2).
Phosphate modulates receptor sulfotyrosine recognition by the chemokine monocyte chemoattractant protein-1 (MCP-1/CCL2).
Organic & biomolecular chemistry (2014-12-24)
Justin P Ludeman, Mahdieh Nazari-Robati, Brendan L Wilkinson, Cheng Huang, Richard J Payne, Martin J Stone
ABSTRACT
Tyrosine sulfation is a widespread post-translational modification that mediates the interactions of secreted and membrane-associated proteins in such varied biological processes as peptide hormone action, adhesion, blood coagulation, complement activation and regulation of leukocyte trafficking. Due to the heterogeneous nature of tyrosine sulfation, detailed biochemical and biophysical studies of tyrosine sulfation rely on homogenous, synthetic sulfopeptides. Here we describe the synthesis of a fluorescent sulfopeptide (FL-R2D) derived from the chemokine receptor CCR2 and the application of FL-R2D in direct and competitive fluorescence anisotropy assays that enable the efficient measurement of binding affinities between sulfopeptides and their binding proteins. Using these assays, we have found that the binding of the chemokine monocyte chemoattractant protein-1 (MCP-1) to sulfated peptides derived from the chemokine receptor CCR2 is highly dependent on the assay buffer. In particular, phosphate buffer at close to physiological concentrations competes with the receptor sulfopeptide by binding to the sulfopeptide binding pocket on the chemokine surface. Thus, physiological phosphate may modulate the receptor binding selectivity of chemokines.
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
MOPS, BioUltra, Molecular Biology, ≥99.5% (titration)
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
BIS-TRIS propane, ≥99.0% (titration)
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
BIS-TRIS propane, BioPerformance Certified, suitable for cell culture, ≥99.0%
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
PIPES, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%