Merck
  • Cyclosporine A induces connective tissue growth factor expression in human gingival fibroblasts: suppression by epigallocatechin-3-gallate.
Cyclosporine A induces connective tissue growth factor expression in human gingival fibroblasts: suppression by epigallocatechin-3-gallate.
Journal of the Formosan Medical Association = Taiwan yi zhi (2014-02-25)
King-Jean Wu, Guay-Fen Huang, Chun-Hao Chen, Hao-Hueng Chang, Yi-Ting Deng
ABSTRACT
Transforming growth factor-β (TGF-β) plays an important role in the pathogenesis of cyclosporine A (CsA)-induced gingival overgrowth (GO). Connective tissue growth factor (CTGF/CCN2) acts as a cofactor with TGF-β to induce the maximal profibrotic effects of TGF-β. We investigated the effects of CsA on CCN2 expression in human gingival fibroblasts (HGFs) and the potential chemopreventive agent for CsA-induced GO. Western blot analyses were used to examine the signaling pathways of CsA-induced CCN2 expression in HGFs and whether epigallocatechin-3-gallate (EGCG), curcumin, or lovastatin can inhibit CsA-induced CCN2 expression. CsA significantly stimulated CCN2 synthesis in HGFs. This effect can be inhibited by c-Jun NH(2)-terminal kinase (JNK) and Smad3 inhibitors but not by TGF-β neutralizing antibody and TGF-β type I receptor inhibitor. Furthermore, EGCG completely blocked CsA-induced CCN2 expression. CsA-induced CCN2 protein expression is mediated through JNK and Smad signaling. CsA may contribute to the pathogenesis of GO through upregulation of CCN2 expression in HGFs. EGCG could be an adjuvant for the prevention of CsA-induced GO.
Product Description
Supelco
(−)-Epigallocatechin gallate, analytical standard
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Epigallocatechin gallate, primary reference standard
USP
Curcumin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Curcumin, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
PD 98,059, solid
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
SIS3, ≥98% (HPLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Supelco
Curcumin, analytical standard
Curcumin, primary reference standard
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O