Merck
  • Apoptotic effect of cisplatin and cordycepin on OC3 human oral cancer cells.
Apoptotic effect of cisplatin and cordycepin on OC3 human oral cancer cells.
Chinese journal of integrative medicine (2013-04-02)
Ying-hui Chen, Lyh-Jyh Hao, Chih-peng Hung, Jung-wei Chen, Sew-fen Leu, Bu-miin Huang
ABSTRACT
To evaluate apoptotic effects of cisplatin and cordycepin as single agent or in combination with cytotoxicity in oral cancer cells. The influences of cisplatin (2.5 μg/mL) and/or cordycepin treatment (10 or 100 μmol/L) to human OC3 oral cancer cell line were investigated by morphological observation for cell death appearance, methylthiazoletetrazolium (MTT) assay for cell viability, flow cytometry assay for cell apoptosis, and Western blotting for apoptotic protein expressions. Data demonstrated that co-administration of cisplatin (2.5 μg/mL) and cordycepin (10 or 100 μmol/L) resulted in the enhancement of OC3 cell apoptosis compared to cisplatin or cordycepin alone treatment (24 h), respectively (P <0.05). In flow cytometry assay, percentage of cells arrested at subG1 phase with co-treatment of cordycepin and cisplatin (30%) was significantly higher than cisplatin (5%) or cordycepin (12%) alone group (P <0.05), confirming a synergistically apoptotic effect of cordycepin and cisplatin. In cellular mechanism study, co-treatment of cordycepin and cisplatin induced more stress-activated protein kinase/Jun terminal kinase (JNK), the expressions of caspase-7, and the cleavage of poly ADP-ribose polymerase (PARP) as compared to cisplatin or cordycepin alone treatment (P <0.05). Cisplatin and cordycepin possess synergistically apoptotic effect through the activation of JNK/caspase-7/PARP pathway in human OC3 oral cancer cell line.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%