Merck
  • Effects of supra-zero storage on human ovarian cortex prior to vitrification-warming.
Effects of supra-zero storage on human ovarian cortex prior to vitrification-warming.
Reproductive biomedicine online (2014-06-24)
Silke Klocke, Claudia Tappehorn, Georg Griesinger
ABSTRACT
This study investigated the effects of supra-zero storage on ovarian cortex of 12 premenopausal patients before cryopreservation. Fresh ovarian tissue (control) was either vitrified immediately or stored at 4°C for 24 h or 48 h before vitrification. This study assessed malondialdehyde release during storage and the capacity to synthesize oestradiol in culture, follicle morphology and the proportion of apoptotic follicles subsequent to vitrification-warming. The malondialdehyde concentration in the storage medium increased significantly in a time-dependent manner (P = 0.029). Oestradiol release during tissue culture decreased statistically significantly in both storage groups compared with the immediate vitrification group (P = 0.043). The proportion of high-quality follicles was significantly different between the fresh group and the two storage groups (24 h, P < 0.01; 48 h, P < 0.001). The proportion of apoptotic follicles was significantly different between the fresh and immediate vitrification groups (P < 0.05). The 24-h and 48-h groups were not different compared with the other groups. This study provides evidence that storage at supra-zero conditions has detrimental effects on the human ovarian cortex. Storage duration should therefore be limited to a minimum to prevent potential damage.
Product Description
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture