Merck
  • The inhibitory effect of angiotensin II on BKCa channels in podocytes via oxidative stress.
The inhibitory effect of angiotensin II on BKCa channels in podocytes via oxidative stress.
Molecular and cellular biochemistry (2014-09-23)
Na Gao, Hui Wang, Xiaochen Zhang, Zhuo Yang
ABSTRACT
Angiotensin II (Ang II) is an important active substance of the renin-angiotensin system (RAS). The present study has confirmed that abnormalities of Ang II may be related with cerebrovascular diseases, endocrine diseases, cardiovascular diseases, liver diseases, such as: cerebral hypoxia, diabetes, obesity, atrial fibrillation, and liver cirrhosis. However, understanding effects of Ang II on podocytes is not enough. This study was to investigate the effects of oxidative stress on the large conductance, Ca(2+)-activated K(+) channels (BKCa). Results from the 3-(4, 5-dimethylthiazol-2-yl)-2, 5-diphenyltetrazolium bromide (MTT) assay showed that Ang II induced podocyte death in a concentration-dependent manner. The measurement of superoxide dismutase (SOD) generation demonstrated that Ang II decreased the total SOD of cellular levels. Meaningfully, pretreatment of a type of ROS scavenger formulations named N-(mercaptopropionyl)-glycine (N-MPG) could inhibit podocyte apoptosis induced by Ang II. Meanwhile, patch-clamp technique was used in this study to detect the effects of Ang II on currents of BKCa channel in podocytes. The results indicated that Ang II inhibited the current amplitude of BKCa channel and decreased the slope of I-V curve. Ang II also made the activation curves of BKCa channel shift to the left. These results may provide a theoretical basis for potential treatment of chronic glomerular disease in the future.
Product Description
Glycine, European Pharmacopoeia (EP) Reference Standard
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, 99%, FCC
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
SAFC
Glycine
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method