Merck
  • Magnesium modulates the expression levels of calcification-associated factors to inhibit calcification in a time-dependent manner.
Magnesium modulates the expression levels of calcification-associated factors to inhibit calcification in a time-dependent manner.
Experimental and therapeutic medicine (2015-02-11)
Jinsheng Xu, Yaling Bai, Jingjing Jin, Junxia Zhang, Shenglei Zhang, Liwen Cui, Huiran Zhang
ABSTRACT
Vascular calcification, a common complication in patients with chronic kidney disease, involves a variety of mechanisms associated with the regulation of calcification-associated factors. Previous clinical studies have indicated that magnesium is involved in the reduction of vascular calcification; however, the mechanism underlying this process remains unknown. The aim of the present study was to investigate the effects of magnesium on β-glycerophosphate (β-GP)-induced calcification and the underlying mechanisms. Primary rat vascular smooth muscle cells (VSMCs) were exposed to 10 mM β-GP in medium with or without the addition of 3 mM magnesium or 2-aminoethoxy-diphenylborate (2-APB; an inhibitor of magnesium transport), for a 14-day period. Calcium deposition and alkaline phosphatase (ALP) activity were measured by Alizarin red staining, quantification of calcium and enzyme-linked immunosorbent assay. The expression levels of core-binding factor α-1 (Cbfα1), matrix Gla protein (MGP) and osteopontin (OPN) were determined by reverse transcription-polymerase chain reaction or western blot analysis, following incubation for 0, 3, 6, 10 and 14 days with the different media. VSMC calcification and ALP activity was reduced significantly in the high-magnesium medium compared with the calcification medium, during the 14-day incubation. The magnesium-induced changes in the VSMCs included a β-GP-induced downregulation of Cbfα1 by day 3 of incubation, an effect that was gradually enhanced over the 14-day period. By contrast, magnesium produced notable increases in MGP and OPN expression levels, with an opposite pattern to that observed in the Cbfα1 expression levels. However, the addition of 2-APB appeared to inhibit the protective effect of magnesium on the VSMCs. Therefore, magnesium was able to effectively reduce β-GP-induced calcification in rat VSMCs by regulating the expression levels of calcification-associated factors in a time-dependent manner.
Product Description
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Glycine, European Pharmacopoeia (EP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Chloral hydrate, crystallized, ≥98.0% (T)
Sigma-Aldrich
Chloral hydrate, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%