Merck
  • Alendronate-induced atypical bone fracture: evidence that the drug inhibits osteogenesis.
Alendronate-induced atypical bone fracture: evidence that the drug inhibits osteogenesis.
Journal of clinical pharmacy and therapeutics (2014-03-26)
S Patntirapong, W Singhatanadgit, S Arphavasin
ABSTRACT
Alendronate (ALN) is used for the treatment of post-menopausal osteoporosis. By reducing bone turnover, it increases bone mineral density. However, recent reports suggest an increased risk of atypical bone fractures after long-term ALN administration. Despite its well-known anti-osteoclastic activity, it is unclear whether ALN also suppresses human mesenchymal stem cell (hMSC)-mediated osteogenesis, thus possibly resulting in atypical bone fragility. We hypothesized that ALN does this and we look at its in vitro effects on osteogenesis. Morphological analysis, reverse transcriptase polymerase chain reaction, cell viability, alkaline phosphatase (ALP) activity and mineralization assays were investigated in hMSCs treated with a wide range of ALN. After treatment with high concentrations of ALN for 3 and 7 days, cell viability was significantly reduced and cell morphology was altered. Osteogenic differentiation of hMSCs was also substantially suppressed as demonstrated by decreased ALP activity although ALN did not affect osteogenic-related genes tested. Furthermore, ALN at all concentrations tested drastically inhibited alizarin red S-positive mineralized matrix. ALN has a strong inhibitory effect on hMSC-mediated osteogenesis by suppressing cell proliferation, osteoblast differentiation and function. The insight gained may help in the development of safer alternatives.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
SAFC
Glycine
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Cetylpyridinium chloride, meets USP testing specifications
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.