Merck
  • Abiogenic Syntheses of Lipoamino Acids and Lipopeptides and their Prebiotic Significance.
Abiogenic Syntheses of Lipoamino Acids and Lipopeptides and their Prebiotic Significance.
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life (2015-08-08)
Gordon Sproul
ABSTRACT
Researchers have formed peptide bonds under a variety of presumed prebiotic conditions. Here it is proposed that these same conditions would have also formed amide bonds between fatty acids and amino acids, producing phosphate-free amphipathic lipoamino acids and lipopeptides. These compounds are known to form vesicles and are ubiquitous in living organisms. They could represent molecules that provided protection by membranes as well as possibilities for proto-life metabolism . It is here demonstrated that when a fatty acid is heated with various amino acids, optimally in the presence of suitable salts or minerals, lipoamino acids are formed. Magnesium and potassium carbonates as well as iron (II) sulfide are found to be particularly useful in these reactions. In this manner N-lauroylglycine, N-lauroylalanine, N-stearoylalanine and several other lipoamino acids have been synthesized. Similarly, when glycylglycine was heated with lauric acid in the presence of magnesium carbonate, the lipopeptide N-lauroylglycylglycine was formed. Such compounds are proposed to have been critical precursors to the development of life.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Magnesium carbonate, meets USP testing specifications
Sigma-Aldrich
Gly-Gly, BioPerformance Certified, suitable for cell culture, ≥99%
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Gly-Gly, ≥99% (titration)
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
SAFC
Glycine
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethyl laurate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl laurate, ≥98%, FCC, FG
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Oxalyl chloride, reagent grade, 98%
Sigma-Aldrich
Oxalyl chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Sodium sulfide hydrate, ≥60%, scales