Merck
  • Characterization of a Synthetic Steroid 24-keto-cholest-5-en-3β, 19-diol as a Neuroprotectant.
Characterization of a Synthetic Steroid 24-keto-cholest-5-en-3β, 19-diol as a Neuroprotectant.
CNS neuroscience & therapeutics (2015-02-14)
Min Yan, Ai-Ling Liu, Shu-Jia Zhou, Li-Peng Tang, Yan-Qiu Ou, Wei Yin, Xin-Ying Chen, Xing-Wen Su, Peng-Xin Qiu, Yi-Jun Huang, Jing-Xia Zhang, Guang-Mei Yan, Tian-Dong Leng
ABSTRACT
Neuroactive steroids represent promising candidates for the treatment of neurological disorders. Our previous studies identified an endogenous steroid cholestane-3β, 5α, 6β-triol (Triol) as a novel neuroprotectant. We aimed to identify a potent candidate for stroke treatment through a screening of Triol analogs. Hypoxia- and glutamate-induced neuronal injury models in vitro, middle cerebral artery occlusion (MCAO)-induced cerebral ischemia model in vivo, fluorescein diacetate (FDA) for alive and propidium iodide (PI) for dead staining, LDH assay, and calcium imaging techniques were used. 24-keto-cholest-5-en-3β, 19-diol (Diol) showed the most potent neuroprotective effect among the screened structurally related compounds. FDA and PI staining showed that Diol concentration dependently increased the survival rate of cerebellar granule neurons (CGNs) challenged with glutamate or hypoxia, with an effective threshold concentration of 2.5 μM. Consistently, the quantitative LDH release assay showed the same concentration-dependent protection in both models. Diol, at 10 μM, potently decreased glutamate- and hypoxia-induced LDH release from 51.6 to 18.2% and 62.1 to 21.7%, respectively, which values are close to the normal LDH release (~16-18%). Moreover, we found Diol effectively decreased MCAO-induced infarction volume in mice from ~23% to 7%, at a dose of 6 mg/kg. We further explored the underlying mechanism and found that Diol attenuated NMDA-induced intracellular calcium ([Ca(2+) ]i ) increase in cortical neurons, suggesting a negative modulatory effect on NMDA receptor. Taken together, we identified Diol as a potent neuroprotectant. It may represent a novel and promising neuroprotectant for stroke intervention.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
SAFC
Glycine
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%