Merck
  • An In Vivo Method to Identify microRNA Targets Not Predicted by Computation Algorithms: p21 Targeting by miR-92a in Cancer.
An In Vivo Method to Identify microRNA Targets Not Predicted by Computation Algorithms: p21 Targeting by miR-92a in Cancer.
Cancer research (2015-06-13)
Xiaoping Su, Huaming Wang, Wei Ge, Mingjin Yang, Jin Hou, Taoyong Chen, Nan Li, Xuetao Cao
ABSTRACT
microRNA (miRNA) dysregulation is involved in the development and progression of various human cancers, including hepatocellular carcinoma (HCC). However, how to identify the miRNAs targeting specific mRNA in cells is a significant challenge because of the interaction complexity and the limited knowledge of rules governing these processes. Some miRNAs are not predictable by current computer algorithms available. Here, using p21 mRNA as target, we established a new method, called miRNA in vivo precipitation (miRIP), to identify which kind of miRNAs can actually bind to the specific mRNA in cells. Several unpredictable miRNAs that bound p21 mRNA in HepG2 and PC-3 cells were identified by the miRIP method. Among these miRNAs identified by miRIP, miR-92a was found and confirmed to interact robustly with p21 mRNA, both in HepG2 and PC-3 cells. miR-92a was found to be remarkably increased in HCC tissues, and higher expression of miR-92a significantly correlated with lower expression of p21, which is related to poor survival of HCC patients. Moreover, inhibition of miR-92a could significantly suppress HCC growth in vitro and in vivo by upregulating p21. Together, miR-92a, which is identified by miRIP, is functionally shown to be associated with HCC growth as an oncogenic miRNA by inhibiting expression of targeting gene p21. In addition, several unpredictable miRNAs that target STAT3 mRNA were also identified by the miRIP method in HepG2 cells. Our results demonstrated that the miRIP approach can effectively identify the unpredictable but intracellular existing miRNAs that target specific mRNA in vivo.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
SAFC
Glycine
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)