Merck
  • Involvement of resveratrol in crosstalk between adipokine adiponectin and hepatokine fetuin-A in vivo and in vitro.
Involvement of resveratrol in crosstalk between adipokine adiponectin and hepatokine fetuin-A in vivo and in vitro.
The Journal of nutritional biochemistry (2015-08-19)
Hee Jae Lee, Yunsook Lim, Soo Jin Yang
ABSTRACT
Metabolic homeostasis is maintained by the coordinated regulation of several physiological processes and organ crosstalk. Especially, the interaction between adipose tissue and liver is critical for the regulation of glucose and lipid metabolism. This study investigated the involvement of resveratrol (RSV) in the crosstalk between adipokine adiponectin and hepatokine fetuin-A. Adipocytes-hepatocytes co-culture system and a high-fat (HF) diet-induced obesity (DIO) mouse model were utilized. Protein levels of adiponectin and fetuin-A were analyzed in adipocytes and hepatocytes with the knockdown of adiponectin and fetuin-A, respectively. After six weeks of the HF diet treatment, RSV was delivered via an osmotic pump for four weeks. The experimental groups were lean control fed with a standard diet, HF diet-induced obese control and HF_RSV (8 mg/kg/day). After 4 weeks of each treatment, blood and tissues were collected, and the levels of adiponectin and fetuin-A were analyzed. RNA interference during co-culture of adipocytes and hepatocytes demonstrated the existence of crosstalk between adiponectin and fetuin-A. The four-week RSV treatment resulted in increased serum adiponectin and decreased serum fetuin-A in diet-induced obesity mice. The serum levels of adiponectin and fetuin-A were inversely related. In epididymal fat depots, RSV increased adiponectin, peroxisome proliferator-activated receptor (PPAR) alpha, PPAR gamma, sirtuin1 and AMP-activated protein kinase (AMPK). RSV lowered fetuin-A and NF-κB, and increased liver AMPK. These results demonstrate the crosstalk between adiponectin and fetuin-A, and suggest that RSV may be involved in adipose tissue and liver crosstalk through the interaction between adiponectin and fetuin-A.
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis