Merck
  • Region-specific alterations of AMPA receptor phosphorylation and signaling pathways in the pilocarpine model of epilepsy.
Region-specific alterations of AMPA receptor phosphorylation and signaling pathways in the pilocarpine model of epilepsy.
Neurochemistry international (2015-05-20)
Mark William Lopes, Samantha Cristiane Lopes, Ana Paula Costa, Filipe Marques Gonçalves, Débora Kurrle Rieger, Tanara Vieira Peres, Helena Eyng, Rui Daniel Prediger, Alexandre Paim Diaz, Jean Costa Nunes, Roger Walz, Rodrigo Bainy Leal
ABSTRACT
Disturbances in glutamatergic transmission and signaling pathways have been associated with temporal lobe epilepsy (TLE) in humans. However, the profile of these alterations within specific regions of the hippocampus and cerebral cortex has not yet been examined. The pilocarpine model in rodents reproduces the main features of TLE in humans. The present study aims to characterize specific alterations of the glutamatergic transmission and signaling pathways in the dorsal (DH) and ventral hippocampus (VH) and temporal cortex (Ctx) of male adult Wistar rats 60 days after pilocarpine treatment (chronic period). The western blotting analyzes show a decrease of AMPA glutamate receptor subunit (GluA1)-Ser(845) phosphorylation; reduction of ERK1 and PKA activity; up-regulation of GFAP and down-regulation of the glutamate transporter EAAT2 expression in the DH. In contrast, in the VH it was observed a decrease of GluA1-Ser(831) phosphorylation and JNKp54 and PKC activity. In the Ctx, only ERK1 phosphorylation/activity decreased. The level of GluA1-Ser(845) phosphorylation and PKA activity (DH) and the level of GluA1-Ser(831) phosphorylation and PKC activity (VH) appear to be correlated, respectively. These findings suggest a differential imbalance of the signaling pathways involved in the site-specific phosphorylation of AMPA receptor in the hippocampus. Furthermore, we suggest that dorsal hippocampus is probably more susceptible to the impairment of glutamate uptake and gliose, since only this area displayed a significant decrease of EAAT2 and increment of GFAP. Taken together, our study suggests that specific neurochemical alterations take place in hippocampal sub regions. This approach may be valuable for understanding the onset of seizures and the alterations of neuronal excitability in specific regions and may help to establish therapeutic targets for treatment of this neuropathology.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
SAFC
Glycine
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))