Merck
  • Altered post-mortem metabolism identified in very fast chilled lamb M. longissimus thoracis et lumborum using metabolomic analysis.
Altered post-mortem metabolism identified in very fast chilled lamb M. longissimus thoracis et lumborum using metabolomic analysis.
Meat science (2015-07-15)
Robyn D Warner, Robin H Jacob, Katja Rosenvold, Simone Rochfort, Craige Trenerry, Tim Plozza, Matthew B McDonagh
ABSTRACT
The aim of this experiment was to use metabolomic techniques to investigate the energy metabolism in lamb M. longissimus thoracis et lumborum subjected to very fast chilling (VFC) post-mortem. The tissue was prepared by 2 different operators and subjected to very fast chilling (less than 0°C within 1.5h of slaughter) or typical chilling regimes (Control; 0°C within 22h of slaughter). Non-targeted metabolomic analysis ((1)H NMR) and targeted analysis ((31)P NMR, HPLC-PDA and HPLC-MS/MS) were used to examine the change in muscle metabolites post-mortem. One VFC treatment, which resulted in a colder core temperature and more tender meat, had higher levels of glycolytic intermediate metabolites pre-rigor as well as more of the end-products of adenosine and nicotine nucleotide metabolism pre-rigor, relative to conventionally chilled treatments. In conclusion, VFC to less than 0°C within 1.5h of slaughter causes considerable changes in metabolism and rigor onset, which are associated with tender meat.
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, 97%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, anhydrous, free-flowing, Redi-Dri, 97%