Merck
Cas9-Guide RNA Directed Genome Editing in Soybean.
Plant physiology (2015-08-22)
Zhongsen Li, Zhan-Bin Liu, Aiqiu Xing, Bryan P Moon, Jessica P Koellhoffer, Lingxia Huang, R Timothy Ward, Elizabeth Clifton, S Carl Falco, A Mark Cigan
ABSTRACT
Recently discovered bacteria and archaea adaptive immune system consisting of clustered regularly interspaced short palindromic repeats (CRISPR) and CRISPR-associated (Cas) endonuclease has been explored in targeted genome editing in different species. Streptococcus pyogenes Cas9-guide RNA (gRNA) was successfully applied to generate targeted mutagenesis, gene integration, and gene editing in soybean (Glycine max). Two genomic sites, DD20 and DD43 on chromosome 4, were mutagenized with frequencies of 59% and 76%, respectively. Sequencing randomly selected transgenic events confirmed that the genome modifications were specific to the Cas9-gRNA cleavage sites and consisted of small deletions or insertions. Targeted gene integrations through homology-directed recombination were detected by border-specific polymerase chain reaction analysis for both sites at callus stage, and one DD43 homology-directed recombination event was transmitted to T1 generation. T1 progenies of the integration event segregated according to Mendelian laws and clean homozygous T1 plants with the donor gene precisely inserted at the DD43 target site were obtained. The Cas9-gRNA system was also successfully applied to make a directed P178S mutation of acetolactate synthase1 gene through in planta gene editing.
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SAFC
Glycine
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)