Merck
  • Application of integrated comprehensive/multidimensional gas chromatography with mass spectrometry and olfactometry for aroma analysis in wine and coffee.
Application of integrated comprehensive/multidimensional gas chromatography with mass spectrometry and olfactometry for aroma analysis in wine and coffee.
Food chemistry (2015-05-09)
Sung-Tong Chin, Graham T Eyres, Philip J Marriott
ABSTRACT
Component coelution in chromatographic analysis complicates identification and attribution of individual odour-active volatile molecules in complex multi-component samples. An integrated system incorporating comprehensive two-dimensional gas chromatography (GC × GC) and multidimensional gas chromatography (MDGC), with flame ionisation, olfactometry and mass spectrometry detection was developed to circumvent data correlation across different systems. Identification of potent odorants in Shiraz wine and the headspace of ground coffee are demonstrated as selected applications. Multiple solid-phase microextraction (SPME) sampling with GC-O located odour-active regions; GC × GC established the complexity of odour-active regions; MDGC provided high-resolution separation for each region; simultaneous 'O' and MS detection completed the analysis for target resolved peaks. Seven odour regions in Shiraz were analysed with MDGC-O/MS detection, revealing 11 odour volatiles through matching of mass spectrometry and retention indices from both separating dimensions, including acetic acid; octen-3-ol; ethyl octanoate; methyl-2-oxo-nonanoate; butanoic acid, 2-methylbutanoic acid, and 3-methylbutanoic acid; 3-(methylthio)-1-propanol; hexanoic acid; β-damascenone; and ethyl-3-phenylpropanoate. A capsicum odour in ground coffee was identified as 2-methoxy-3-isobutylpyrazine with a 5-fold increase in S/N of the odorant when acquired using a 6-time cumulative SPME sampling approach.
Sigma-Aldrich
Butyric acid, ≥99%, FG
Sigma-Aldrich
(±)-2-Methylbutyric acid, natural, ≥98%, FG
Sigma-Aldrich
Pyridinium dichromate, 98%
Sigma-Aldrich
Butyric acid, ≥99%
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 4
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #5, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 4
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis