Merck
Cyclotron production of (44)Sc: From bench to bedside.
Nuclear medicine and biology (2015-06-22)
Nicholas P van der Meulen, Maruta Bunka, Katharina A Domnanich, Cristina Müller, Stephanie Haller, Christiaan Vermeulen, Andreas Türler, Roger Schibli
ABSTRACT
(44)Sc, a PET radionuclide, has promising decay characteristics (T1/2 = 3.97 h, Eβ(+)av = 632 keV) for nuclear imaging and is an attractive alternative to the short-lived (68)Ga (T1/2 = 68 min, Eβ(+)av = 830 keV). The aim of this study was the optimization of the (44)Sc production process at an accelerator, allowing its use for preclinical and clinical PET imaging. (44)CaCO3 targets were prepared and irradiated with protons (~11 MeV) at a beam current of 50 μA for 90 min. (44)Sc was separated from its target material using DGA extraction resin and concentrated using SCX cation exchange resin. Radiolabeling experiments at activities up to 500 MBq and stability tests were performed with DOTANOC by investigating different scavengers, including gentisic acid. Dynamic PET of an AR42J tumor-bearing mouse was performed after injection of (44)Sc-DOTANOC. The optimized chemical separation method yielded up to 2 GBq (44)Sc of high radionuclidic purity. In the presence of gentisic acid, radiolabeling of (44)Sc with DOTANOC was achieved with a radiochemical yield of ~99% at high specific activity (10 MBq/nmol) and quantities which would allow clinical application. The dynamic PET images visualized increasing uptake of (44)Sc-DOTANOC into AR42J tumors and excretion of radioactivity through the kidneys of the investigated mouse. The concept "from-bench-to-bedside" was clearly demonstrated in this extended study using cyclotron-produced (44)Sc. Sufficiently high activities of (44)Sc of excellent radionuclidic purity are obtainable for clinical application, by irradiation of enriched calcium at a cyclotron. This work demonstrates a promising basis for introducing (44)Sc to clinical routine of nuclear imaging using PET.
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Tantalum(V) ethoxide, packaged for use in deposition systems
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF