Merck
  • Three-dimensional constructs using hyaluronan cell carrier as a tool for the study of cancer stem cells.
Three-dimensional constructs using hyaluronan cell carrier as a tool for the study of cancer stem cells.
Journal of biomedical materials research. Part B, Applied biomaterials (2014-10-29)
Cristina Martínez-Ramos, Myriam Lebourg
ABSTRACT
Cancer research focuses increasingly on cancer stem cell study as those cells are thought to be the root of chemo and radioresistance of the most aggressive cancer types. Nevertheless, two-dimensional (2D) cell culture and even three-dimensional (3D) spheroid models, with their limited ability to reflect cell-extracellular matrix interactions, are not ideal for the study of cancer stem cells (CSCs). In this study, we establish a 3D in vitro cancer model using a synthetic and natural scaffold with tunable features and show that U87 cells cultured in this system acquire a stem-cell like phenotype. U87 astrocytoma cells were grown on polycaprolactone (PCL)-2D flat substrates (2D) and PCL-3D scaffolds (3D) eventually containing hyaluronic acid (3D-HA). Cell viability, growth patterns, morphology, and cell surface marker expression (CD44, RHAMM and CD133) were studied to assess the effect of 3D culture and presence of HA. 3D scaffold, but most prominently presence of HA induced changes in cell morphology and marker expression; 3D-HA cultures showed features of aggregates; moreover, markedly increased expression of Nestin, CD44, RHAMM, and CD133 in 3D-HA scaffolds were found. the behavior of U87 in our 3D-HA model is more similar to tumor growth in vivo and a stem-like phenotype is promoted. Thus, the 3D-HA scaffold could provide a useful model for CSCs study and anti-cancer therapeutics research in vitro and may have preclinical application for the screening of drug candidates.
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Tyramine hydrochloride, ≥98%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
SAFC
Glycine
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)