Merck
  • Synergistic interactions among flavonoids and acetogenins in Graviola (Annona muricata) leaves confer protection against prostate cancer.
Synergistic interactions among flavonoids and acetogenins in Graviola (Annona muricata) leaves confer protection against prostate cancer.
Carcinogenesis (2015-04-12)
Chunhua Yang, Sushma Reddy Gundala, Rao Mukkavilli, Subrahmanyam Vangala, Michelle D Reid, Ritu Aneja
ABSTRACT
Phytochemical complexity of plant extracts may offer health-promoting benefits including chemotherapeutic and chemopreventive effects. Isolation of 'most-active fraction' or single constituents from whole extracts may not only compromise the therapeutic efficacy but also render toxicity, thus emphasizing the importance of preserving the natural composition of whole extracts. The leaves of Annona muricata, commonly known as Graviola, are known to be rich in flavonoids, isoquinoline alkaloids and annonaceous acetogenins. Here, we demonstrate phytochemical synergy among the constituents of Graviola leaf extract (GLE) compared to its flavonoid-enriched (FEF) and acetogenin-enriched (AEF) fractions. Comparative quantitation of flavonoids revealed enrichment of rutin (~7-fold) and quercetin-3-glucoside (Q-3-G, ~3-fold) in FEF compared to GLE. In vivo pharmacokinetics and in vitro absorption kinetics of flavonoids revealed enhanced bioavailability of rutin in FEF compared to GLE. However, GLE was more effective in inhibiting in vitro prostate cancer proliferation, viability and clonogenic capacity compared to FEF. Oral administration of 100mg/kg bw GLE showed ~1.2-fold higher tumor growth-inhibitory efficacy than FEF in human prostate tumor xenografts although the concentration of rutin and Q-3-G was more in FEF. Contrarily, AEF, despite its superior in vitro and in vivo efficacy, resulted in death of the mice due to toxicity. Our data indicate that despite lower absorption and bioavailability of rutin, maximum efficacy was achieved in the case of GLE, which also comprises of other phytochemical groups including acetogenins that make up its natural complex environment. Hence, our study emphasizes on evaluating the nature of interactions among Graviola leaf phytochemcials for developing favorable dose regimen for prostate cancer management to achieve optimal therapeutic benefits.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent