Merck
  • Structure-function dissection of Myxococcus xanthus CarD N-terminal domain, a defining member of the CarD_CdnL_TRCF family of RNA polymerase interacting proteins.
Structure-function dissection of Myxococcus xanthus CarD N-terminal domain, a defining member of the CarD_CdnL_TRCF family of RNA polymerase interacting proteins.
PloS one (2015-03-27)
Diego Bernal-Bernal, Aránzazu Gallego-García, Gema García-Martínez, Francisco García-Heras, María Angeles Jiménez, S Padmanabhan, Montserrat Elías-Arnanz
ABSTRACT
Two prototypes of the large CarD_CdnL_TRCF family of bacterial RNA polymerase (RNAP)-binding proteins, Myxococcus xanthus CarD and CdnL, have distinct functions whose molecular basis remain elusive. CarD, a global regulator linked to the action of several extracytoplasmic function (ECF) σ-factors, binds to the RNAP β subunit (RNAP-β) and to protein CarG via an N-terminal domain, CarDNt, and to DNA via an intrinsically unfolded C-terminal domain resembling eukaryotic high-mobility-group A (HMGA) proteins. CdnL, a CarDNt-like protein that is essential for cell viability, is implicated in σA-dependent rRNA promoter activation and interacts with RNAP-β but not with CarG. While the HMGA-like domain of CarD by itself is inactive, we find that CarDNt has low but observable ability to activate ECF σ-dependent promoters in vivo, indicating that the C-terminal DNA-binding domain is required to maximize activity. Our structure-function dissection of CarDNt reveals an N-terminal, five-stranded β -sheet Tudor-like domain, CarD1-72, whose structure and contacts with RNAP-β mimic those of CdnL. Intriguingly, and in marked contrast to CdnL, CarD mutations that disrupt its interaction with RNAP-β did not annul activity. Our data suggest that the CarDNt C-terminal segment, CarD61-179, may be structurally distinct from its CdnL counterpart, and that it houses at least two distinct and crucial function determinants: (a) CarG-binding, which is specific to CarD; and (b) a basic residue stretch, which is also conserved and functionally required in CdnL. This study highlights the evolution of shared and divergent interactions in similar protein modules that enable the distinct activities of two related members of a functionally important and widespread bacterial protein family.
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)