Merck
  • The influence of lignin on steam pretreatment and mechanical pulping of poplar to achieve high sugar recovery and ease of enzymatic hydrolysis.
The influence of lignin on steam pretreatment and mechanical pulping of poplar to achieve high sugar recovery and ease of enzymatic hydrolysis.
Bioresource technology (2015-09-24)
Richard P Chandra, QiuLu Chu, Jinguang Hu, Na Zhong, Mandy Lin, Jin-Suk Lee, Jack Saddler
ABSTRACT
With the goal of enhancing overall carbohydrate recovery and reducing enzyme loading refiner mechanical pulping and steam pretreatment (210°C, 5 min) were used to pretreat poplar wood chips. Neutral sulphonation post-treatment indicated that, although the lignin present in the steam pretreated substrate was less reactive, the cellulose-rich, water insoluble component was more accessible to cellulases and Simons stain. This was likely due to lignin relocation as the relative surface lignin measured by X-ray photoelectron spectroscopy increased from 0.4 to 0.8. The integration of sulphite directly into steam pretreatment resulted in the solubilisation of 60% of the lignin while more than 80% of the carbohydrate present in the original substrate was recovered in the water insoluble fraction after Na2CO3 addition. More than 80% of the sugars present in the original cellulose and xylan could be recovered after 48 h using an enzyme loading of 20 mg protein/g cellulose at a 10% substrate concentration.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium sulfite, BioUltra, anhydrous, ≥98% (RT)
Sigma-Aldrich
Sodium sulfite, BioXtra, ≥98%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium sulfite, ≥98%
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium sulfite, puriss. p.a., reag. Ph. Eur., anhydrous, 98-100.0% (iodometric)
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium sulfite, ACS reagent, ≥98.0%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Sodium sulfite, anhydrous, Redi-Dri, reagent grade, ≥98%
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Sodium chlorite, puriss. p.a., 80% (RT)