Merck
  • Spectrin alpha is important for rear polarization of the microtubule organizing center during migration and spindle pole assembly during division of neointimal smooth muscle cells.
Spectrin alpha is important for rear polarization of the microtubule organizing center during migration and spindle pole assembly during division of neointimal smooth muscle cells.
Cytoskeleton (Hoboken, N.J.) (2015-04-24)
Rosalind V Silverman-Gavrila, Lorelei B Silverman-Gavrila, Khawaja Hasan Bilal, Michelle P Bendeck
ABSTRACT
Directed migration of smooth muscle cells (SMCs) from the media to the intima and their subsequent proliferation are key events in atherosclerosis as these cells contribute to the bulk and stability of atheromatous plaques. We showed previously that two cytoskeleton-associated proteins, RHAMM and ARPC5, play important roles in rear polarization of the microtubule organizing centre (MTOC), directed migration, and in maintaining cell division fidelity. These proteins were analyzed to predict additional potential interacting partners using the bioinformatics programs BLAST, ClustalW, and PPI Spider. We identified spectrin alpha, a protein with a known role in actin polymerization as part of the pathway. We show that in migrating SMCs spectrin alpha localizes at the nodes of the actin net, and it partially colocalizes with RHAMM in the perinuclear region. In dividing SMCs spectrin alpha is present at spindle poles and midbody. Moreover, we show that spectrin alpha and RHAMM interact in a complex. Using siRNA to knockdown spectrin disrupted SMC migration, MTOC polarization, and the assembly of a polygonal actin net dorsolateral of the nucleus. Spectrin alpha knockdown also disrupted the organization of the bipolar spindle, chromosome division, and cytokinesis during cell division. The identification of interacting partners such as spectrin alpha and the decoding of pathways involved in polarity regulation during the migration of smooth muscle cells in atherosclerosis is important for identifying atherosclerosis biomarkers and developing therapeutic agents to block atherosclerotic plaque formation.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
MISSION® esiRNA, targeting human SMC1B
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
MISSION® esiRNA, targeting human DYM
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Dym
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
SAFC
Glycine
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)