Skip to Content
Merck
Get up to 22% off for Pi Day until 3/26.Save Now

BDPH9BCE052D

Bide Pharmatech

Methyl 3-bromo-6-chloropyrazine-2-carboxylate

Synonym(s):

methyl 3-bromo-6-chloro-2-pyrazinecarboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCE052D is not currently sold in your country.

About This Item

Linear Formula:
C6H4O2N2Cl1Br1
CAS Number:
Molecular Weight:
251.47
MDL number:
Product BDPH9BCE052D is not currently sold in your country.

Inchi Code

1S/C6H4BrClN2O2/c1-12-6(11)4-5(7)9-2-3(8)10-4/h2H,1H3

Inchi Key

NJAYXNFNZKQGNE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or Semi-solid or liquid or lump

Looking for similar products? Visit Product Comparison Guide

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library